Learn More
HELZ2 Polyclonal Antibody, Invitrogen™
Description
Specifications
Specifications
| Antigen | HELZ2 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | Dulbecco′s PBS with 150mM NaCl, 50% glycerol and 0.02% sodium azide; pH 7.4 |
| Gene | HELZ2 |
| Gene Accession No. | Q9BYK8 |
| Gene Alias | PDIP1-alpha |
| Gene Symbols | HELZ2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.