Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Tocris Bioscience™ HEMADO
GREENER_CHOICE

Catalog No. p-4783513
Click to view available options
Quantity:
10 mg
50 mg
This item is not returnable. View return policy
This item is not returnable. View return policy

High affinity selective A3 agonist

High affinity and selective adenosine A3 receptor agonist (Ki values are 1.1, 327, 1230 and > 30,000 nM for human A3, A1, A2A and A2B receptors respectively).

Chemical Identifiers

CAS 403842-38-6
Molecular Formula C17H23N5O4

Specifications

Quantity 10 mg
Formula Weight 361.4
Purity >99%
Molecular Weight (g/mol) 361.402
Chemical Name or Material HEMADO
Recommended Storage Desiccate at +4°C
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.