Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HEMK2 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
Antigen | HEMK2 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15648620
![]() |
Novus Biologicals
NBP15648620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP156486
![]() |
Novus Biologicals
NBP156486 |
100 μL |
Each for $487.50
|
|
|||||
Description
HEMK2 Polyclonal specifically detects HEMK2 in Human, Mouse samples. It is validated for Western Blot.Specifications
HEMK2 | |
Polyclonal | |
Rabbit | |
Q96F73 | |
29104 | |
Synthetic peptides corresponding to N6AMT1(N-6 adenine-specific DNA methyltransferase 1 (putative)) The peptide sequence was selected from the N terminal of N6AMT1. Peptide sequence MAGENFATPFHGHVGRGAFSDVYEPAEDTFLLLNALEAAAAELAGVEICL. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
C21orf127, chromosome 21 open reading frame 127, EC 2.1.1.-, HemK methyltransferase family member 2, HEMK2, M.HsaHemK2P, MGC19995, MTQ2, N(6)-adenine-specific DNA methyltransferase 1, N-6 adenine-specific DNA methyltransferase 1 (putative), N6AMT, N6-DNA-methyltransferase, PRED28 | |
N6AMT1 | |
IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title