Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemoglobin A1c Antibody (Hb4), Novus Biologicals™
SDP

Catalog No. NBP105486 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105486 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105486 Supplier Novus Biologicals Supplier No. NBP105486
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hemoglobin A1c Monoclonal specifically detects Hemoglobin A1c in Human samples. It is validated for Immunoassay.

Specifications

Antigen Hemoglobin A1c
Applications Immunoassay
Classification Monoclonal
Clone Hb4
Conjugate Unconjugated
Dilution Immunoassay
Gene Alias alpha one globin, alpha-1 globin, alpha-1-globin, alpha-globin, CD31, hemoglobin alpha 1 globin chain, Hemoglobin alpha chain, hemoglobin alpha-1 chain, hemoglobin subunit alpha, hemoglobin, alpha 1, MGC126895, MGC126897
Gene Symbols HBA1
Host Species Mouse
Immunogen Hemoglobin human HbA10 (Hb4)
Purification Method Protein A purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 3039
Test Specificity Hemoglobin human HbA10 (Hb4)
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.