Learn More
Description
Specifications
Specifications
| Antigen | Hemoglobin alpha |
| Applications | ELISA, Immunoprecipitation, Radioimmune Assays (RIA) |
| Classification | Monoclonal |
| Clone | 9A5 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | ELISA 1:100 - 1:2000, Immunoprecipitation 1:10 - 1:500, Radioimmunoassay 1:100 - 1:2000 |
| Gene Accession No. | P69905 |
| Gene Alias | alpha one globin, alpha-1 globin, alpha-1-globin, alpha-globin, CD31, hemoglobin alpha 1 globin chain, Hemoglobin alpha chain, hemoglobin alpha-1 chain, hemoglobin subunit alpha, hemoglobin, alpha 1, MGC126895, MGC126897 |
| Gene Symbols | HBA1 |
| Show More |
For Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
