Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemoglobin alpha Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB11041080 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11041080 1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11041080 Supplier Novus Biologicals Supplier No. NB11041080
Add to Cart
Add to Cart

Goat Polyclonal Antibody

Hemoglobin alpha Polyclonal specifically detects Hemoglobin alpha in Human samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Hemoglobin alpha
Applications Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate HRP
Dilution Western Blot 1:1000-1:30000, ELISA 1:10000-1:100000, Immunohistochemistry 1:200-1:500, Immunocytochemistry/Immunofluorescence 1:200-1:500, Immunohistochemistry-Paraffin 1:200 - 1:500
Formulation PBS and 0.2% BSA with 0.05% Pro-Clean 400
Gene Accession No. P69905
Gene Alias alpha one globin, alpha-1 globin, alpha-1-globin, alpha-globin, CD31, hemoglobin alpha 1 globin chain, Hemoglobin alpha chain, hemoglobin alpha-1 chain, hemoglobin subunit alpha, hemoglobin, alpha 1, MGC126895, MGC126897
Gene Symbols HBA1
Host Species Goat
Immunogen Human Hemoglobin
Purification Method Affinity Purified
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 3039
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.