Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemoglobin Antibody (2A1), mFluor Violet 610 SE, Novus Biologicals™
SDP

Catalog No. NB340472 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB340472 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB340472 Supplier Novus Biologicals Supplier No. NBP320120MFV610
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Hemoglobin Monoclonal antibody specifically detects Hemoglobin in Mouse samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence

Specifications

Antigen Hemoglobin
Applications Western Blot, ELISA, Immunocytochemistry
Classification Monoclonal
Clone 2A1
Conjugate mFluor Violet 610 SE
Dilution Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
Formulation 50mM Sodium Borate
Gene Alias alpha one globin;alpha-1 globin;alpha-1-globin;alpha-2 globin chain;Alpha-globin;ALPHA-GLOBIN GENE;delta globin;Haemoglobin;Hb;HbA1c;hemoglobin alpha 1 globin chain;Hemoglobin alpha chain;Hemoglobin subunit alpha;hemoglobin, alpha 1
Host Species Rabbit
Immunogen Murine hybridomas were generated using spleen cells from aging (8-10 month old) NZB/W F1 animals. Hybridomas secreting antibodies reactive to murine hemoglobin were subcloned by limiting dilution and antibody isotypes determined using commercial reagents. Eight-week-old NZB/W F1 were immunized subcutaneously with 50 mg mHb antigen emulsified in Complete Fruend's Adjuvant.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.