Learn More
Invitrogen™ Hemoglobin Monoclonal Antibody (HB11-203.1), Invitrogen™
Description
MA1-10807 detects Hemoglobin from human samples. This antibody does not detect bovine, chicken, equine, ovine, or porcine Hemoglobin. MA1-10807 has been successfully used in ELISA and Western blot applications. The MA1-10807 immunogen is purified human hemoglobin.
Specifications
Specifications
| Antigen | Hemoglobin |
| Applications | ELISA, Western Blot |
| Classification | Monoclonal |
| Clone | HB11-203.1 |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with no preservative; pH 7.4 |
| Gene | HBA1 |
| Gene Accession No. | P68871, P69905 |
| Gene Alias | alpha one globin; alpha-1 globin; alpha-1-globin; alpha-2 globin chain; Alpha-globin; ALPHA-GLOBIN GENE; delta globin; Haemoglobin; Hb; HBA1; HbA1c; HBA2; HBA-T3; HBH; hemoglobin alpha 1 globin chain; Hemoglobin alpha chain; Hemoglobin subunit alpha; hemoglobin subunit alpha 1; hemoglobin, alpha 1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.