Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemoglobin zeta Antibody (GT1544), Novus Biologicals™
SDP

Catalog No. NB177230 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177230 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177230 Supplier Novus Biologicals Supplier No. NBP313543
Add to Cart
Add to Cart
This item is not returnable. View return policy

Mouse Monoclonal Antibody

Hemoglobin zeta Monoclonal antibody specifically detects Hemoglobin zeta in Human samples. It is validated for Western Blot, Immunofluorescence

Specifications

Antigen Hemoglobin zeta
Applications Western Blot, Immunofluorescence
Classification Monoclonal
Clone GT1544
Conjugate Unconjugated
Formulation PBS
Gene Alias HBAZ, HBZ2, hemoglobin subunit zeta, Hemoglobin zeta chain, hemoglobin, zeta, zeta-globin
Host Species Mouse
Immunogen Recombinant protein encompassing a sequence within the center region of human Hemoglobin zeta. The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Research Discipline Immunology
Primary or Secondary Primary
Gene ID (Entrez) 3050
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.