Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemoglobin gamma A Rabbit anti-Human, Rat, Clone: JM84-10, Novus Biologicals™
SDP

Catalog No. NBP266885 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP266885 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP266885 Supplier Novus Biologicals Supplier No. NBP266885
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

HBG1/2 Monoclonal specifically detects HBG1/2 in Human, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen HemoglobingammaA
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone JM84-10
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, Immunohistochemistry 1:10 - 1:500, Immunocytochemistry/Immunofluorescence 1:50-1:200, Immunohistochemistry-Paraffin 1:50-1:200
Formulation TBS (pH7.4), 0.05% BSA, 40% Glycerol with 0.05% Sodium Azide
Gene Alias A-gamma globin, gamma A hemoglobin, gamma globin, gamma-1-globin, Hb F Agamma, HBGA, HBGR, Hemoglobin gamma-1 chain, Hemoglobin gamma-A chain, hemoglobin subunit gamma-1, hemoglobin, gamma A, hemoglobin, gamma, regulator of, HSGGL1
Gene Symbols HBG1
Host Species Rabbit
Immunogen Synthetic peptide within Human HBG1/2 aa 2-45/ 147. (SwissProt: P69891 Human; SwissProt: P69892 Human)
Purification Method Protein A purified
Quantity 100 μL
Research Discipline Cardiovascular Biology
Primary or Secondary Primary
Gene ID (Entrez) 3047
Target Species Human, Rat
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.