Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hemopexin Rabbit anti-Human, Clone: 36, Novus Biologicals™
SDP

Catalog No. NB020856 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
50 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB020856 50 μg
NB020855 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB020856 Supplier Novus Biologicals Supplier No. NBP28968650UL
Add to Cart
Add to Cart

Rabbit Monoclonal Antibody

Hemopexin Monoclonal specifically detects Hemopexin in Human samples. It is validated for ELISA.

Specifications

Antigen Hemopexin
Applications ELISA
Classification Monoclonal
Clone 36
Conjugate Unconjugated
Dilution ELISA 1:5000-1:10000
Formulation 0.2 um filtered solution in PBS
Gene Alias beta-1B-glycoprotein, FLJ56652, hemopexin, HX
Host Species Rabbit
Immunogen Recombinant Human Hemopexin protein
Purification Method Protein A purified
Quantity 50 μg
Regulatory Status RUO
Research Discipline Angiogenesis, Cancer, Endothelial Cell Markers, Hematopoietic Stem Cell Markers, HIF Target Genes, Hypoxia, Stem Cell Markers, Tyrosine Kinases
Primary or Secondary Primary
Gene ID (Entrez) 3263
Target Species Human
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.