Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HEPACAM2 Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB342664 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB342664 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB342664 Supplier Novus Biologicals Supplier No. NBP281720PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HEPACAM2 Polyclonal antibody specifically detects HEPACAM2 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA

Specifications

Antigen HEPACAM2
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot, ELISA
Formulation PBS
Gene Alias HEPACAM family member 2, MIKI
Host Species Rabbit
Immunogen Rabbit polyclonal HEPACAM2 antibody was raised against a 19 amino acid peptide near the carboxy terminus of human HEPACAM2. The immunogen is located within amino acids 370 - 420 of HEPACAM2. Amino Acid Squence: CVIKQKLEGRPETEYRKAQT
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cancer, Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 253012
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.