Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heparan Sulfate Proteoglycan 2 Antibody (A74) - BSA Free, Novus Biologicals™
SDP

Catalog No. NBP22352301 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP22352301 0.1 mL
NB019315 0.025 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NBP22352301 Supplier Novus Biologicals Supplier No. NBP2235230.1ML
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Heparan Sulfate Proteoglycan 2 Monoclonal specifically detects Heparan Sulfate Proteoglycan 2 in Human, Bovine samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, Immunoaffinity Purification.

Specifications

Antigen Heparan Sulfate Proteoglycan 2
Applications Western Blot, ELISA, Immunohistochemistry, Immunoprecipitation
Classification Monoclonal
Clone A74
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000, Immunohistochemistry 1:10-1:500, Immunoprecipitation 1:10-1:500
Formulation 10mM Phosphate (pH 7.4) and 0.5M NaCl with 15mM Sodium Azide
Gene Alias basement membrane-specific heparan sulfate proteoglycan core protein, endorepellin (domain V region), heparan sulfate proteoglycan 2, HSPG, perlecan, perlecan proteoglycan, PLCSchwartz-Jampel syndrome 1 (chondrodystrophic myotonia), PRCAN, SJA, SJS, SJS1
Gene Symbols HSPG2
Host Species Mouse
Immunogen Bovine corneal endothelial cells
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 3339
Test Specificity Clone A74 is highly specific for perlecan. There is no evidence for cross-reactivity with other connective tissue proteins (vitronectin, fibronectin, elastin, collagen, laminin). Clone A74 cross-reacts with human perlecan. Other species have not been tested.
Target Species Human, Bovine
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.