Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatic Sinusoidal Endothelial Cells Antibody (SE-1), Novus Biologicals™
SDP

Catalog No. NB11068095T Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11068095T 0.025 mL
NB11068095 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB11068095T Supplier Novus Biologicals Supplier No. NB11068095SS
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatic Sinusoidal Endothelial Cells Monoclonal specifically detects Hepatic Sinusoidal Endothelial Cells in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Hepatic Sinusoidal Endothelial Cells
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Monoclonal
Clone SE-1
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1-5 ug/ml, Flow Cytometry reported in scientific literature (PMID 9428229), Immunohistochemistry 1-5 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-5 ug/ml, Immunohistochemistry-Frozen 1-5 ug/ml
Gene Alias Hepatic cells, Hepatic Sinusoidal Endothelial Cells, HSE, SE1, Sinusoidal Endothelial Cells
Host Species Mouse
Immunogen Rat Hepatic Sinusoidal Endothelial Cells
Purification Method Protein A purified
Quantity 0.025 mL
Regulatory Status RUO
Research Discipline Endothelial Cell Markers
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.