Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatic Sinusoidal Endothelial Cells Antibody (SE-1), Novus Biologicals™
SDP

Mouse Monoclonal Antibody

$207.00 - $489.50

Specifications

Antigen Hepatic Sinusoidal Endothelial Cells
Clone SE-1
Concentration 1 mg/mL
Dilution Western Blot 1-5 ug/ml, Flow Cytometry reported in scientific literature (PMID 9428229), Immunohistochemistry 1-5 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-5 ug/ml, Immunohistochemistry-Frozen 1-5 ug/ml
Applications Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
NB11068095T
SDP
View Documents
Novus Biologicals
NB11068095SS
0.025 mL
Each for $207.00
Only null left
Add to Cart
 
NB11068095
SDP
View Documents
Novus Biologicals
NB11068095
0.1 mL
Each for $489.50
Only null left
Add to Cart
 
Description

Description

Hepatic Sinusoidal Endothelial Cells Monoclonal specifically detects Hepatic Sinusoidal Endothelial Cells in Human, Mouse, Rat samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.
Specifications

Specifications

Hepatic Sinusoidal Endothelial Cells
1 mg/mL
Western Blot, Flow Cytometry, Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Unconjugated
Mouse
Endothelial Cell Markers
Hepatic cells, Hepatic Sinusoidal Endothelial Cells, HSE, SE1, Sinusoidal Endothelial Cells
IgG2a κ
Protein A purified
SE-1
Western Blot 1-5 ug/ml, Flow Cytometry reported in scientific literature (PMID 9428229), Immunohistochemistry 1-5 ug/ml, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin 1-5 ug/ml, Immunohistochemistry-Frozen 1-5 ug/ml
Monoclonal
Purified
RUO
Human, Mouse, Rat
Rat Hepatic Sinusoidal Endothelial Cells
Primary
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.