Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatic Sinusoidal Endothelial Cells Antibody (SE-1), PE, Novus Biologicals™
SDP

Catalog No. NB11068095Q Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB11068095Q 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB11068095Q Supplier Novus Biologicals Supplier No. NB11068095PE
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatic Sinusoidal Endothelial Cells Monoclonal specifically detects Hepatic Sinusoidal Endothelial Cells in Human, Mouse, Rat samples. It is validated for Flow Cytometry.

Specifications

Antigen Hepatic Sinusoidal Endothelial Cells
Applications Flow Cytometry
Classification Monoclonal
Clone SE-1
Conjugate PE
Dilution Flow Cytometry
Gene Alias Hepatic cells, Hepatic Sinusoidal Endothelial Cells, HSE, SE1, Sinusoidal Endothelial Cells
Host Species Mouse
Immunogen Rat Hepatic Sinusoidal Endothelial Cells
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Mouse, Rat
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.