Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis A Surface Antigen Antibody (BGN/B5B3), Biotin, Novus Biologicals™
SDP

Catalog No. NB10062197B Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10062197B 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB10062197B Supplier Novus Biologicals Supplier No. NB10062197B
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatitis A Surface Antigen Monoclonal specifically detects Hepatitis A Surface Antigen in Virus samples. It is validated for ELISA, Radioimmune Assays (RIA).

Specifications

Antigen Hepatitis A Surface Antigen
Applications ELISA, Radioimmune Assays (RIA)
Classification Monoclonal
Clone BGN/B5B3
Conjugate Biotin
Dilution ELISA
Formulation PBS with 0.05% Sodium Azide
Host Species Mouse
Immunogen Lysate from foetal kidney cells of rhesus monkey infected with extract from patient with active viral Hepatitis A.
Purification Method Protein G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-62197 is specific for Hepatitis A surface antigen.
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.