Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B preS2 Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB344949 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB344949 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB344949 Supplier Novus Biologicals Supplier No. NB10093580MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hepatitis B preS2 Polyclonal antibody specifically detects Hepatitis B preS2 in Human samples. It is validated for Western Blot

Specifications

Antigen Hepatitis B preS2
Applications Western Blot
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot
Formulation 50mM Sodium Borate
Gene Alias HBV pre S2 Antigen, HBV pre S2 protein, Hepatitis B Virus pre S2 protein, PreS2/S antigen, S
Host Species Rabbit
Immunogen Synthetic peptide within residues 130-150 of the 42 kDa preS2 HBV protein. This sequence represents a B cell epitope.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.