Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis B Virus Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NB100664300.1ML
Description
Hepatitis B Virus Polyclonal specifically detects Hepatitis B Virus in Virus samples. It is validated for Western Blot, ELISA.Specifications
| Hepatitis B Virus | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.2) with 0.1% Sodium Azide | |
| Rabbit | |
| Protein G purified | |
| RUO | |
| Recognises the 39 and 41 kD (L-protein) in Blotting. Both bands are peptide blockable. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot, ELISA | |
| 5.0 mg/mL | |
| Western Blot 1:250-1:500, ELISA 1:100-1:2000 | |
| HBsAg, Hepatitis B Receptor Binding Fragment Pre S | |
| Synthetic peptide corresponding to the amino acids 21-47 of the native molecule conjugated to Keyhole limpet Haemocyanin. | |
| 0.1 mL | |
| Primary | |
| Virus | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction