Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Hepatitis B Virus Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Specifications
| Antigen | Hepatitis B Virus |
|---|---|
| Concentration | 5.0 mg/mL |
| Dilution | Western Blot 1:250-1:500, ELISA 1:100-1:2000 |
| Applications | Western Blot, ELISA |
| Classification | Polyclonal |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NB017773
![]() |
Novus Biologicals
NB100664300.1ML |
0.1 mL | N/A | N/A | N/A | ||||
NB10066430
![]() |
Novus Biologicals
NB10066430 |
0.5 mL | N/A | N/A | N/A | ||||
Description
Hepatitis B Virus Polyclonal specifically detects Hepatitis B Virus in Virus samples. It is validated for Western Blot, ELISA.Specifications
| Hepatitis B Virus | |
| Western Blot 1:250-1:500, ELISA 1:100-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.2) with 0.1% Sodium Azide | |
| Synthetic peptide corresponding to the amino acids 21-47 of the native molecule conjugated to Keyhole limpet Haemocyanin. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
| 5.0 mg/mL | |
| Western Blot, ELISA | |
| Unconjugated | |
| Rabbit | |
| Virus | |
| HBsAg, Hepatitis B Receptor Binding Fragment Pre S | |
| IgG | |
| Protein G purified | |
| Recognises the 39 and 41 kD (L-protein) in Blotting. Both bands are peptide blockable. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title