Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus Antibody, Novus Biologicals™
SDP

Rabbit Polyclonal Antibody

Supplier:  Novus Biologicals NB10066430

 View more versions of this product

Catalog No. NB10066430


Description

Description

Hepatitis B Virus Polyclonal specifically detects Hepatitis B Virus in Virus samples. It is validated for Western Blot, ELISA.
Specifications

Specifications

Hepatitis B Virus
Polyclonal
Unconjugated
PBS (pH 7.2) with 0.1% Sodium Azide
Rabbit
Protein G purified
RUO
Recognises the 39 and 41 kD (L-protein) in Blotting. Both bands are peptide blockable.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
IgG
Western Blot, ELISA
5.0 mg/mL
Western Blot 1:250-1:500, ELISA 1:100-1:2000
HBsAg, Hepatitis B Receptor Binding Fragment Pre S
Synthetic peptide corresponding to the amino acids 21-47 of the native molecule conjugated to Keyhole limpet Haemocyanin.
0.5 mL
Primary
Virus
Purified
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.

For Research Use Only