Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus X antigen Antibody (X36C), DyLight 350, Novus Biologicals™
SDP

Catalog No. NBP222445UV Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP222445UV 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP222445UV Supplier Novus Biologicals Supplier No. NBP222445UV
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatitis B Virus X antigen Monoclonal specifically detects Hepatitis B Virus X antigen in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Hepatitis B Virus X antigen
Applications Western Blot, ELISA
Classification Monoclonal
Clone X36C
Conjugate DyLight 350
Dilution Western Blot, ELISA
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HBVgp3 X protein HBx, Hepatitis B Virus X Protein, Peptide X, pX, X protein
Gene Symbols HBVgp3
Host Species Mouse
Immunogen Baculovirus expressed recombinant HBx.
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 944566
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.