Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis B Virus X antigen Antibody (X36C), mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB355690 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB355690 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB355690 Supplier Novus Biologicals Supplier No. NBP222445MFV500
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatitis B Virus X antigen Monoclonal antibody specifically detects Hepatitis B Virus X antigen in Virus samples. It is validated for Western Blot, ELISA, Immunoprecipitation

Specifications

Antigen Hepatitis B Virus X antigen
Applications Western Blot, ELISA, Immunoprecipitation
Classification Monoclonal
Clone X36C
Conjugate mFluor Violet 500 SE
Dilution Western Blot, ELISA, Immunoprecipitation
Formulation 50mM Sodium Borate
Gene Alias HBVgp3 X protein, HBx, Hepatitis B Virus X Protein, Peptide X, pX, X protein
Host Species Mouse
Immunogen Baculovirus expressed recombinant HBx.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 944566
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.