Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C Virus Antibody, PerCP, Novus Biologicals™
SDP

Catalog No. NB360593 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB360593 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB360593 Supplier Novus Biologicals Supplier No. NBP227586PCP
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hepatitis C Virus Polyclonal antibody specifically detects Hepatitis C Virus in Virus samples. It is validated for Western Blot

Specifications

Antigen Hepatitis C Virus
Applications Western Blot
Classification Polyclonal
Conjugate PerCP
Dilution Western Blot
Formulation PBS
Gene Alias HCV NS5, Hepatitis C virus non structural protein 5, Hepatitis C virus nonstructural protein 5, Hepatitis C Virus NS5
Host Species Rabbit
Immunogen Reacts with residues 33-43 [CGVYLLPRRGPR] of HCV core, env, and part of E2/NS1 of HCV.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.