Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C Virus Core Antigen Antibody (C7-50), HRP, Novus Biologicals™
SDP

Catalog No. NB211442 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB211442 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB211442 Supplier Novus Biologicals Supplier No. NB1202740H
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatitis C Virus Core Antigen Monoclonal antibody specifically detects Hepatitis C Virus Core Antigen in Human, Mouse, Virus samples. It is validated for Western Blot, ELISA, Immunohistochemistry

Specifications

Antigen Hepatitis C Virus Core Antigen
Applications Western Blot, ELISA, Immunohistochemistry
Classification Monoclonal
Clone C7-50
Conjugate HRP
Formulation PBS
Gene Alias Core protein p19, HCcAg, HCV core antigen, HCV core protein
Host Species Mouse
Immunogen Purified HCV core-GST fusion protein (genotype 1b).
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Infections (Virus Bacteria and Parasites), Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 951475
Target Species Human, Mouse, Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.