Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis C Virus Core Antigen Antibody (C7-50), PE/Cy5.5, Novus Biologicals™
SDP

Catalog No. N1202740PC5 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N1202740PC5 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N1202740PC5 Supplier Novus Biologicals Supplier No. NB1202740PECY55
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatitis C Virus Core Antigen Monoclonal specifically detects Hepatitis C Virus Core Antigen in Human, Mouse, Virus samples. It is validated for Flow Cytometry.

Specifications

Antigen Hepatitis C Virus Core Antigen
Applications Flow Cytometry
Classification Monoclonal
Clone C7-50
Conjugate PE-Cyanine5.5
Dilution Flow Cytometry
Formulation PBS with 0.05% Sodium Azide
Gene Alias Core protein p19, HCcAg, HCV core antigen, HCV core protein
Gene Symbols POLY
Host Species Mouse
Immunogen Purified HCV core-GST fusion protein (genotype 1b).
Purification Method Affinity Purified
Quantity 0.1 mL
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Gene ID (Entrez) 951475.0
Test Specificity Detects hepatitis C virus (HCV) core protein from transfected human and primate cell lines.
Target Species Human, Mouse, Virus
Content And Storage Store at 4°C in the dark. Do not freeze.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.