Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis D Virus Large Delta Antibody (HL1051), Novus Biologicals™
SDP

Catalog No. NB177389 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB177389 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. NB177389 Supplier Novus Biologicals Supplier No. NBP313702
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Monoclonal Antibody

Hepatitis D Virus Large Delta Monoclonal antibody specifically detects Hepatitis D Virus Large Delta in Virus samples. It is validated for Western Blot

Specifications

Antigen Hepatitis D Virus Large Delta
Applications Western Blot
Classification Monoclonal
Clone HL1051
Conjugate Unconjugated
Formulation PBS
Gene Alias Large delta antigen, Large hepatitis delta antigen, L-HDAg, p27
Host Species Rabbit
Immunogen Carrier-protein conjugated synthetic peptide encompassing a sequence within the C-terminus region of Hepatitis D Virus Large Antigen (Hepatitis delta virus genotype I (isolate D380)). The exact sequence is proprietary.
Purification Method Protein A purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.