Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatitis G Virus Antibody - BSA Free, Novus Biologicals™
SDP

Catalog No. NB017932 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB017932 0.025 mg
NB10093584 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB017932 Supplier Novus Biologicals Supplier No. NB10093584SS
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Hepatitis G Virus Polyclonal specifically detects Hepatitis G Virus in Human, Virus samples. It is validated for Western Blot.

Specifications

Antigen Hepatitis G Virus
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:500-1:1000
Formulation PBS with 0.02% Sodium Azide
Gene Alias Hepatitis GB virus C, HGV
Host Species Rabbit
Immunogen Reacts with residues 2268-2276 [CDKCEARQE] of HGV.
Purification Method Affinity Purified
Quantity 0.025 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity Hepatitis G Virus
Target Species Virus, Human
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.