Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatocyte Specific Antigen Antibody (HSA98) - Azide and BSA Free, Novus Biologicals™
SDP

Catalog No. NB397159 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB397159 0.1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB397159 Supplier Novus Biologicals Supplier No. NBP2346450.1MG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatocyte Specific Antigen Monoclonal antibody specifically detects Hepatocyte Specific Antigen in Human samples. It is validated for ELISA

Specifications

Antigen Hepatocyte Specific Antigen
Applications ELISA
Classification Monoclonal
Clone HSA98
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution ELISA 1 to 5 μg/mL
Formulation 10 mM PBS
Gene Alias Hepatocyte, HSA
Host Species Mouse
Immunogen HEP-3B human hepatocellular carcinoma cells
Purification Method Protein A or G purified
Quantity 0.1 mg
Regulatory Status RUO
Research Discipline Cellular Markers
Primary or Secondary Primary
Target Species Human
Content And Storage Store at -20°C to -80°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2b κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.