Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatocyte Specific Antigen Antibody (OCH1E5), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. NP234699A32 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NP234699A32 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NP234699A32 Supplier Novus Biologicals Supplier No. NBP234699AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatocyte Specific Antigen Monoclonal specifically detects Hepatocyte Specific Antigen in Human, Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Hepatocyte Specific Antigen
Applications Immunohistochemistry, Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OCH1E5
Conjugate Alexa Fluor 532
Dilution Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Hepatocyte, HSA
Host Species Mouse
Immunogen Extract of a formalin-fixed, rejected-allograft of a human liver
Purification Method Protein A or G purified
Quantity 0.1 mL
Primary or Secondary Primary
Target Species Human, Canine
Content And Storage Store at 4°C in the dark.
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.