Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatocyte Specific Antigen Antibody (OCH1E5), DyLight 680, Novus Biologicals™
SDP

Catalog No. NBP234699FR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP234699FR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP234699FR Supplier Novus Biologicals Supplier No. NBP234699FR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatocyte Specific Antigen Monoclonal specifically detects Hepatocyte Specific Antigen in Human, Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Hepatocyte Specific Antigen
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen)
Classification Monoclonal
Clone OCH1E5
Conjugate DyLight 680
Dilution Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen
Gene Alias Hepatocyte, HSA
Host Species Mouse
Immunogen Extract of a formalin-fixed, rejected-allograft of a human liver
Purification Method Protein A or G purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Canine
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1 κ
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.