Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Hepatocyte Specific Antigen Mouse anti-Canine, Human, Clone: OCH1E5, Novus Biologicals™
SDP

Catalog No. NB3433620UG Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μg
20 μg
200 μg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB3433620UG 20 μg
NB34336100U 100 μg
NB34336200U 200 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. NB3433620UG Supplier Novus Biologicals Supplier No. NBP23433620UG
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Hepatocyte Specific Antigen Monoclonal specifically detects Hepatocyte Specific Antigen in Human, Canine samples. It is validated for Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin.

Specifications

Antigen Hepatocyte Specific Antigen
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), SDS-Page
Classification Monoclonal
Clone OCH1E5
Conjugate Unconjugated
Dilution Immunocytochemistry/Immunofluorescence 0.5-1.0ug/ml, Immunohistochemistry-Paraffin 0.5-1ug/ml, Immunohistochemistry-Frozen 0.5-1ug/mlimmunohistochemistry-Paraffin 0.5-1ug/ml, SDS-Page
Formulation PBS with 0.05% BSA. with 0.05% Sodium Azide
Gene Alias Hepatocyte, HSA
Host Species Mouse
Immunogen Extract of a formalin-fixed, rejected-allograft of a human liver
Purification Method Protein G purified
Quantity 20 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Human, Canine
Content And Storage Store at 4C.
Form Purified
Isotype IgG1
Show More Show Less

RUO

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.