Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Heptane CHROMASOLV™, for HPLC, ≥99%, Honeywell Riedel-de Haën™
Shop All Honeywell Research Chemicals Products
Click to view available options
Quantity:
100 mL
1 L
2.5 L
4 x 2.5 L
4 x 4 L
6 x 1 L
Description
- For HPLC and organic synthesis
- High-purity, multipurpose solvents
- High lot-to-lot consistency
Specifications
Specifications
| Melting Point | -91°C |
| Density | 0.684g/mL at 25°C |
| Boiling Point | 98°C |
| Quantity | 4 x 4 L |
| Refractive Index | n20/D 1.387 |
| Linear Formula | CH3(CH2)5CH3 |
| Vapor Pressure | 83mmHg (37.7°C) |
| Beilstein | 1730763 |
| Formula Weight | 100.2g/mol |
| Percent Purity | ≥99% |
| Show More |
Safety and Handling
| Hazard Statement | P210-P273-P301 + P310-P304 + P340 + P312-P331-P391 |
| Precautionary Statement | H225-H304-H315-H336-H410 |
| EINECSNumber | 205-563-8 |
| RTECSNumber | MI7700000 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction