Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Heptyl Methyl Sulfide 97.0+%, TCI America™
SDP

Catalog No. h061310ml Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
10 mL

Chemical Identifiers

CAS 20291-61-6
Molecular Formula C8H18S
Molecular Weight (g/mol) 146.292
MDL Number MFCD00027314
InChI Key FJDWJOQOEZRIDJ-UHFFFAOYSA-N
PubChem CID 44824
IUPAC Name 1-methylsulfanylheptane
SMILES CCCCCCCSC

Specifications

Color Yellow
Boiling Point 195°C
Quantity 10 mL
Formula Weight 146.29
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Heptyl Methyl Sulfide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.