Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERC1 Antibody, Novus Biologicals™
SDP

Catalog No. NBP105968 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105968 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105968 Supplier Novus Biologicals Supplier No. NBP105968
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HERC1 Polyclonal specifically detects HERC1 in Human samples. It is validated for Immunoprecipitation.

Specifications

Antigen HERC1
Applications Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Immunoprecipitation 2 - 5 ug/mg lysate, Western Blot (Negative)
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. NP_003913.3
Gene Alias EC 6.3.2, guanine nucleotide exchange factor p532, hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1(CHC1)-like domain (RLD) 1, hect domain and RCC1-like domain 1, HECT domain and RCC1-like domain-containing protein 1, p532probable E3 ubiquitin-protein ligase HERC1, p619EC 6.3.2.-
Gene Symbols HERC1
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 125 and 175 of human hect (homologous to the E6-AP (UBE3A) carboxyl terminus) domain and RCC1 (CHC1)-like domain (RLD) 1 using the numbering given in entry NP_003913.3 (GeneID 892
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8925
Target Species Human
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.