Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERC2 Antibody, Novus Biologicals™
SDP

Catalog No. NBP105970 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP105970 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP105970 Supplier Novus Biologicals Supplier No. NBP105970
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HERC2 Polyclonal specifically detects HERC2 in Human, Mouse samples. It is validated for Western Blot, Immunoprecipitation.

Specifications

Antigen HERC2
Applications Western Blot, Immunoprecipitation
Classification Polyclonal
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:2000-1:10000, Immunoprecipitation 1:10-1:500
Formulation Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide
Gene Accession No. O95714
Gene Alias D15F37S1, DKFZp547P028, E3 ubiquitin-protein ligase HERC2, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 2, hect domain and RLD 2, jdf2, KIAA0393, p528, probable E3 ubiquitin-protein ligase HERC2, SHEP1
Gene Symbols HERC2
Host Species Rabbit
Immunogen The immunogen recognized by this antibody maps to a region between residue 4784 and 4834 of human hect domain and RLD 2 using the numbering given in entry NP_004658.3 (GeneID 8924).
Molecular Weight of Antigen 527 kDa
Purification Method Affinity Purified
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8924
Test Specificity Based on 100% sequence identity, this antibody is predicted to react with Mouse, Rat, Zebrafish, X. tropicalis, Panda, Rhesus Monkey, Gorilla, Chimpanzee, West Indian ocean coelacanth, Nile tilapia and Spotted green pufferfish.
Target Species Human, Mouse
Content And Storage Store at 4C. Do not freeze.
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.