Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERC5 Rabbit anti-Human, Mouse, Rat, Polyclonal, Novus Biologicals™
SDP

Catalog No. NB094563 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB094563 20 μL
NB094562 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB094563 Supplier Novus Biologicals Supplier No. NBP30498720UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HERC5 Polyclonal specifically detects HERC5 in Human, Mouse, Rat samples. It is validated for Western Blot.

Specifications

Antigen HERC5
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 50% glycerol, pH7.3.
Gene Alias CEB1E3 ISG15--protein ligase HERC5, CEBP1, cyclin-E binding protein 1, Cyclin-E-binding protein 1, EC 6.3.2, EC 6.3.2.-, HECT domain and RCC1-like domain-containing protein 5, hect domain and RLD 5, HECT E3 ubiquitin ligase, probable E3 ubiquitin-protein ligase HERC5
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 360-700 of human HERC5 (NP_057407.2).
Purification Method Affinity purified
Quantity 20 μL
Regulatory Status RUO
Research Discipline Cell Cycle and Replication
Primary or Secondary Primary
Gene ID (Entrez) 51191
Target Species Human, Mouse, Rat
Content And Storage Store at -20°C. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.