Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
HERC6 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $501.50
Specifications
Antigen | HERC6 |
---|---|
Applications | Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Host Species | Rabbit |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP15502520
![]() |
Novus Biologicals
NBP15502520UL |
20 μL |
Each for $206.00
|
|
|||||
NBP155025
![]() |
Novus Biologicals
NBP155025 |
100 μL |
Each for $501.50
|
|
|||||
Description
HERC6 Polyclonal specifically detects HERC6 in Human samples. It is validated for Western Blot.Specifications
HERC6 | |
Polyclonal | |
Rabbit | |
Q8IVU3 | |
55008 | |
Synthetic peptides corresponding to HERC6(hect domain and RLD 6) Antibody(against the N terminal of HERC6. Peptide sequence LSKDSQVFSWGKNSHGQLGLGKEFPSQASPQRVRSLEGIPLAQVAAGGAH. | |
Primary |
Western Blot | |
Unconjugated | |
RUO | |
EC 6.3.2, EC 6.3.2.-, FLJ20637, HECT domain and RCC1-like domain-containing protein 6, hect domain and RLD 6, potential ubiquitin ligase, probable E3 ubiquitin-protein ligase HERC6 | |
HERC6 | |
IgG | |
115 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title