Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 Antibody, HRP, Novus Biologicals™
SDP

Catalog No. NB212027 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB212027 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB212027 Supplier Novus Biologicals Supplier No. NB600516H
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Herpes Simplex Virus 1 Polyclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Western Blot, ELISA

Specifications

Antigen Herpes Simplex Virus 1
Applications Western Blot, ELISA
Classification Polyclonal
Conjugate HRP
Formulation PBS
Gene Alias Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1
Host Species Rabbit
Immunogen Rabbits were immunized with Herpes Simplex Virus 1 (HSV1) (SQPELAPEDPED) conjugated to KLH. Antibody was isolated by affinity chromatography using the peptide immobilized on solid support.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.