Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 Antibody, Novus Biologicals™
SDP

Catalog No. NB018100 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
0.5 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB018100 0.1 mL
NB1209533 0.5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB018100 Supplier Novus Biologicals Supplier No. NB12095330.1ML
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Herpes Simplex Virus 1 Polyclonal specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen.

Specifications

Antigen Herpes Simplex Virus 1
Applications Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), Immunohistochemistry
Classification Polyclonal
Conjugate Unconjugated
Dilution Immunohistochemistry 1:10-1:500, Immunohistochemistry-Paraffin 1:10-1:500, Immunohistochemistry-Frozen 1:10-1:500
Formulation Citrate, pH 6.0, containing less than 0.1% sodium azide with Sodium Azide
Gene Alias Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1
Host Species Rabbit
Immunogen Whole rabbit corneal cells, infected with herpes simplex virus type I (strain Mac Intyre), were solubilized with detergent.
Purification Method Affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity This antibody reacts with Herpes Simplex Virus 1 (HSV1) specific antigens and with antigens common to Herpes Simplex Virus 1 (HSV1) and II virus. It reacts with all the major glycoproteins present in the viral envelope as well as with at least one core protein.
Target Species Virus
Content And Storage Store at 4°C.
Form Purified
Isotype IgG
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.