Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 Antibody (T111), Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB50476A350 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB50476A350 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB50476A350 Supplier Novus Biologicals Supplier No. NB500476AF350
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Herpes Simplex Virus 1 Monoclonal antibody specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen Herpes Simplex Virus 1
Applications ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone T111
Conjugate Alexa Fluor 350
Dilution ELISA, Immunocytochemistry/Immunofluorescence, Immunocytochemistry
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1
Host Species Mouse
Immunogen Exctract of Herpes Simplex Virus 1 infected VERO (green monkey kidney) cells.
Purification Method Protein A purified
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity Recognizes gB antigen of both Herpes Simplex Virus 1 &2
Target Species Virus
Content And Storage Store at 4°C in the dark.
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.