Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 Antibody (T96), Janelia Fluor™ 646, Novus Biologicals™
SDP

Catalog No. NB500477646 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500477646 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500477646 Supplier Novus Biologicals Supplier No. NB500477JF646
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Herpes Simplex Virus 1 Monoclonal specifically detects Herpes Simplex Virus 1 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HerpesSimplexVirus1
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone T96
Conjugate Janelia Fluor 646
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias Herpes simplex virus, Herpes simplex virus 1, Herpes simplex virus type 1, HSV 1, Human herpesvirus 1, Human herpesvirus type 1
Host Species Mouse
Immunogen Exctract of HSV-1 infected VERO (green monkey kidney) cells.
Purification Method Ion exchange chromatography
Quantity 0.1 mL
Primary or Secondary Primary
Test Specificity The antibody T96 recognizes gC antigen of HSV1 (Herpes Simplex Virus type 1), a dsDNA virus, member of Herpesviridae family.
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.