Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 1 ICP8 Antibody (10A3), mFluor Violet 450 SE, Novus Biologicals™
SDP

Catalog No. NB310330 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB310330 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB310330 Supplier Novus Biologicals Supplier No. NB1002770MFV450
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Herpes Simplex Virus 1 ICP8 Monoclonal specifically detects Herpes Simplex Virus 1 ICP8 in Virus samples. It is validated for Immunohistochemistry, Western Blot.

Specifications

Antigen Herpes Simplex Virus 1 ICP8
Applications Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 10A3
Conjugate mFluor Violet 450 SE
Dilution Western Blot, Immunohistochemistry
Formulation 50mM Sodium Borate
Gene Alias DBP, Herpes simplex virus type 1 DBP, Herpes simplex virus type 1 ICP8, Herpes simplex virus type 1 ICP8 major DNA binding protein, Herpes simplex virus type 1 infected cell protein 8, Herpes simplex virus type 1 major DNA binding protein, Herpes simplex virus type 1 UL29, HSV1 DBP, HSV1 ICP8, HSV1 infected cell protein 8, HSV1 major DNA binding protein, HSV1 UL29, ICP 8, ICP 8 protein, ICP8, Infected cell protein 8, Major DNA binding protein, UL 29, UL29
Host Species Mouse
Immunogen Herpes Simplex Virus 1 ICP8 purified from U-35-VERO cells.
Purification Method Protein A purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.