Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 2 Antibody (HSVA33), Novus Biologicals™
SDP

Catalog No. NB1102515 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.2 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB1102515 0.2 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB1102515 Supplier Novus Biologicals Supplier No. NB1102515
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Herpes Simplex Virus 2 Monoclonal antibody specifically detects Herpes Simplex Virus 2 in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Herpes Simplex Virus 2
Applications Western Blot, ELISA
Classification Monoclonal
Clone HSVA33
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA
Gene Alias Herpes Simplex Virus Type 2
Host Species Mouse
Immunogen Hybridoma clone has been derived from hybridization of Sp2/0 myeloma cells with spleen cells of Balb/c mice immunized with purified HSV from strain BH.
Purification Method Protein G purified
Quantity 0.2 mg
Regulatory Status RUO
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity Reacts with 56-64 kD band corresponding to glycoprotein D band in immunoblotting of purified HSV under reduced conditions and with 120-140 kD band in immunoblotting of HSV-infected Vero cells lysates under non-reduced conditions.
Target Species Virus
Content And Storage Store at 4C. Do not freeze.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.