Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 2 Antibody (T303), DyLight 488, Novus Biologicals™
SDP

Catalog No. NB500478G Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB500478G 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB500478G Supplier Novus Biologicals Supplier No. NB500478G
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Herpes Simplex Virus 2 Monoclonal antibody specifically detects Herpes Simplex Virus 2 in Virus samples. It is validated for Western Blot, ELISA, Immunocytochemistry, Immunofluorescence.

Specifications

Antigen Herpes Simplex Virus 2
Applications Western Blot, ELISA, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone T303
Conjugate DyLight 488
Dilution Western Blot, ELISA, Immunocytochemistry/Immunofluorescence
Gene Alias Herpes Simplex Virus Type 2
Host Species Mouse
Immunogen Exctract of Herpes Simplex Virus 2 infected VERO (green monkey kidney) cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity The antibody T303 recognizes gG antigen of HSV2 (Herpes Simplex Virus type 2), a dsDNA virus, member of Herpesviridae family.
Target Species Virus
Content And Storage Store at 4C in the dark.
Isotype IgG2a
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.