Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus 2 Glycoprotein D Antibody (A33) - BSA Free, Novus Biologicals™
SDP

Catalog No. NB10063170 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.025 mg
0.1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB10063170 0.1 mg
NB017542 0.025 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. NB10063170 Supplier Novus Biologicals Supplier No. NB10063170

Mouse Monoclonal Antibody

Herpes Simplex Virus 2 Glycoprotein D Monoclonal antibody specifically detects Herpes Simplex Virus 2 Glycoprotein D in Virus samples. It is validated for Western Blot, ELISA.

Specifications

Antigen Herpes Simplex Virus 2 Glycoprotein D
Applications Western Blot, ELISA
Classification Monoclonal
Clone A33
Concentration 1.0 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:100-1:2000, ELISA 1:100-1:2000
Gene Alias Glycoprotein D precursor; Herpes Simplex Virus Type 2 Glycoprotein D; HHV2gp69; US6; Virion glycoprotein D
Host Species Mouse
Immunogen Herpes Simplex Virus 2 Glycoprotein D
Purification Method Protein G purified
Quantity 0.1 mg
Regulatory Status RUO
Primary or Secondary Primary
Test Specificity NB100-63170 is specific for Herpes Simplex virus 2 glycoprotein D (HSV2 gD) a major component of the virion envelope and infected cell membranes that is essential for virus entry into cells. This antibody reacts with a 56-64 kD band, corresponding to glycoprotein D, in an HSV (purified) and infected Vero cell lysate immunoblot.
Target Species Virus
Content And Storage Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG1
Show More Show Less

For Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.