Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus UL42 Antibody (2H4), Alexa Fluor™ 532, Novus Biologicals™
SDP

Catalog No. N102762A532 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N102762A532 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. N102762A532 Supplier Novus Biologicals Supplier No. NB1002762AF532
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HSV UL42 Monoclonal specifically detects HSV UL42 in Non-species specific samples. It is validated for ELISA, Immunocytochemistry/Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot.

Specifications

Antigen HSV UL42
Applications ELISA, Immunocytochemistry, Immunofluorescence, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 2H4
Conjugate Alexa Fluor 532
Dilution Western Blot, ELISA, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50 mM sodium borate with 0.05% sodium azide
Gene Alias HHV1gp061
Host Species Mouse
Immunogen Baculovirus-expressed HSV DNA polymerase (POL) and POL/UL42 complex.
Purification Method Protein A purified
Quantity 0.1 mL
Research Discipline Virology Bacteria and Parasites
Primary or Secondary Primary
Test Specificity May recognize UL42 bound to Herpes Simplex Virus polymerase. Recognizes amino acids 360-366 in UL42.
Target Species Non-species specific
Content And Storage Store at 4°C in the dark.
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.