Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Herpes Simplex Virus VP16 Antibody, Alexa Fluor™ 350, Novus Biologicals™
SDP

Catalog No. NB207209 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB207209 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB207209 Supplier Novus Biologicals Supplier No. NB600950AF350
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Herpes Simplex Virus VP16 Polyclonal antibody specifically detects Herpes Simplex Virus VP16 in Virus samples. It is validated for Western Blot

Specifications

Antigen Herpes Simplex Virus VP16
Applications Western Blot
Classification Polyclonal
Conjugate Alexa Fluor 350
Formulation 50mM Sodium Borate
Host Species Rabbit
Immunogen Synthetic peptide corresponding to amino acids 474-487 of the herpes simplex virus Herpes Simplex Virus VP16 protein, conjugated to KLH via an N-terminal added cysteine.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Primary or Secondary Primary
Target Species Virus
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.