Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERPUD1 Antibody (OTI1A2), DyLight 755, Novus Biologicals™
SDP

Catalog No. NBP271492IR Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NBP271492IR 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NBP271492IR Supplier Novus Biologicals Supplier No. NBP271492IR
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

HERPUD1 Monoclonal specifically detects HERPUD1 in Human, Mouse, Rat, Canine, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.

Specifications

Antigen HERPUD1
Applications Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence
Classification Monoclonal
Clone OTI1A2
Conjugate DyLight 755
Dilution Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence
Formulation 50mM Sodium Borate with 0.05% Sodium Azide
Gene Alias HERPMIF1, homocysteine-inducible endoplasmic reticulum stress-inducible ubiquitin-likedomain member 1 protein, homocysteine-inducible, endoplasmic reticulum stress-inducible, ubiquitin-likedomain member 1, homocysteine-responsive endoplasmic reticulum-resident ubiquitin-like domainmember 1 protein, KIAA0025MMS-inducible, Methyl methanesulfonate (MMF)-inducible fragment protein 1, Mif1, SUP
Gene Symbols HERPUD1
Host Species Mouse
Immunogen Full length recombinant protein of human HERPUD1 (NP_001010989) produced in HEK293T cells.
Purification Method Affinity Purified
Quantity 0.1 mL
Primary or Secondary Primary
Gene ID (Entrez) 9709.0
Target Species Human, Mouse, Rat, Canine, Monkey
Content And Storage Store at 4°C in the dark.
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.