Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

HERV Antibody, mFluor Violet 500 SE, Novus Biologicals™
SDP

Catalog No. NB362460 Shop All R&D Systems Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
NB362460 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. NB362460 Supplier Novus Biologicals Supplier No. NB10093579MFV500
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

HERV Polyclonal antibody specifically detects HERV in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Neutralization

Specifications

Antigen HERV
Applications Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin), Immunohistochemistry (Frozen), In vitro Assay, Blocking Assay
Classification Polyclonal
Conjugate mFluor Violet 500 SE
Dilution Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin, Immunohistochemistry-Frozen, In vitro assay, Block/Neutralize
Formulation 50mM Sodium Borate
Gene Alias endogenous retroviral family W, env(C7), member 1, ENV, envelope glycoprotein, Envelope polyprotein gPr73, enverin, Env-W, ENVW, HERV-7q, HERV7Q, HERV-7q Envelope protein, HERV-tryptophan envelope protein, HERV-W, HERV-W Env glycoprotein, HERV-W envelope protein, HERV-W_7q21.2 provirus ancestral Env polyprotein, HERV-W{7q21.1} provirus ancestral Env polyprotein, HERVWENV, HERV-W-ENV, HERVWenvelope protein, human endogenous retrovirus W envC7-1 envelope protein, Syncytin, syncytin A, Syncytin-1
Host Species Rabbit
Immunogen Synthetic peptide corresponding to residues 42-50 [QRPGNIDAPC] of the HERV protein.
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Cell Biology
Primary or Secondary Primary
Gene ID (Entrez) 30816
Target Species Human
Content And Storage Store at 4C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.